Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylbenzonitrile, 97%

$120.23 - $120.23

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylbenzonitrile
CAS 5724-56-1
Molecular Formula C9H9N
Formula Weight 131.18
MDL Number MFCD00019744
Beilstein 1932620
Synonym 3-Cyano-o-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1158906

  SDS
alfa aesar™
A1158906
5g Each for $120.23
Description & Specifications

Specifications

Formula Weight 131.18
Chemical Name or Material 2,3-Dimethylbenzonitrile
Assay 97%
Molecular Formula C9H9N
MDL Number MFCD00019744
CAS 5724-56-1
Beilstein 1932620
Synonym 3-Cyano-o-xylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
RTECS Number DI4357600
Odor Almond-like
Boiling Point 64° to 66°C (0.8mmHg)
Melting Point 23° to 25°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.