Please login to your online account to display your discounted pricing

2,3-Dimethylbenzoic acid, 98%, ACROS Organics™

$33.90 - $33.90

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethylbenzoic acid
CAS 603-79-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O2
Linear Formula (CH3)C6H3COH
Formula Weight 150.18
MDL Number MFCD00002479
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306500050

  SDS
thermo fisher scientific
306500050
Glass bottle 5g Each for $33.90
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 150.18
Quantity 5g
Color Beige to light brown
Melting Point 143° to 147°C
CAS Max % 100.0
Synonym vic.-o-Xylylic acid
Infrared Spectrum Authentic
Linear Formula (CH3)C6H3COH
Chemical Name or Material 2, 3-Dimethylbenzoic acid
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H10O2
CAS 603-79-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00002479
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.