Please login to your online account to display your discounted pricing

2,3-Dimethylbenzoic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  306500050

 View more versions of this product

Catalog No. AC306500050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Quantity 5g
Physical Form Crystalline Powder
Formula Weight 150.18
Color Beige to light brown
Melting Point 143° to 147°C
CAS Max % 100.0
Synonym vic.-o-Xylylic acid
Infrared Spectrum Authentic
Linear Formula (CH3)C6H3COH
Packaging Glass bottle
Assay Percent Range 98%
Chemical Name or Material 2, 3-Dimethylbenzoic acid
CAS 603-79-2
Molecular Formula C9H10O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00002479
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2, 3-Dimethylbenzoic acid
CAS 603-79-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O2
Linear Formula (CH3)C6H3COH
Formula Weight 150.18
MDL Number MFCD00002479
Synonym vic.-o-Xylylic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents