Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylbenzeneboronic acid, 98%

$36.01 - $388.74

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylbenzeneboronic acid
CAS 183158-34-1
Molecular Formula C8H11BO2
Formula Weight 149.99
MDL Number MFCD01863524
Beilstein 8050763
Synonym 2,3-Dimethylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2394206

  SDS
alfa aesar™
B2394206
5g Each for $109.54

AAB2394203

  SDS
alfa aesar™
B2394203
1g Each for $36.01

AAB2394214

  SDS
alfa aesar™
B2394214
25g Each for $388.74
Description & Specifications

Specifications

Chemical Name or Material 2,3-Dimethylbenzeneboronic acid
CAS 183158-34-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C8H11BO2
Melting Point ≈175°C
Formula Weight 149.99
MDL Number MFCD01863524
Beilstein 8050763
Quantity 5g
Synonym 2,3-Dimethylphenylboronic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.