missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dimethylbenzene-1-carbonyl chloride, 97%, Maybridge

Chemical Identifiers

CAS 21900-46-9
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H9ClO
Linear Formula (CH3)2C6H3C(O)Cl
Formula Weight 168.62
Chemical Name or Material 2, 3-Dimethylbenzene-1-carbonyl chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
TL00220EA SDS Fisher Scientific Company
TL00220EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21900-46-9
97%
(CH3)2C6H3C(O)Cl
2, 3-Dimethylbenzene-1-carbonyl chloride
97.0
C9H9ClO
168.62

Specifications

10g
97.0
97%
(CH3)2C6H3C(O)Cl
2, 3-Dimethylbenzene-1-carbonyl chloride
21900-46-9
Amber Glass Bottle
C9H9ClO
168.62

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger