Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylbenzamide, 98%

$24.18 - $375.58

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylbenzamide
CAS 5580-34-7
Molecular Formula C9H11NO
Formula Weight 149.19
MDL Number MFCD00017570
Beilstein 2574226
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1358503

 
alfa aesar™
A1358503
1g Each for $24.18

AAA1358506

 
alfa aesar™
A1358506
5g Each for $88.36

AAA1358514

 
alfa aesar™
A1358514
25g Each for $375.58
Description & Specifications

Specifications

Formula Weight 149.19
Chemical Name or Material 2,3-Dimethylbenzamide
Assay 98%
Molecular Formula C9H11NO
MDL Number MFCD00017570
CAS 5580-34-7
Beilstein 2574226
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 155° to 159°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.