missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylbenzaldehyde 96.0+%, TCI America™
Supplier: TCI America D34635G
Specifications
| 2,3-Dimethylbenzaldehyde | |
| Yellow | |
| C9H10O | |
| 5 g | |
| UIFVCPMLQXKEEU-UHFFFAOYSA-N | |
| 2,3-dimethylbenzaldehyde | |
| 34224 | |
| ≥96.0% (GC) |
| 5779-93-1 | |
| 110°C | |
| MFCD00798004 | |
| benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde | |
| CC1=CC=CC(=C1C)C=O | |
| 134.178 | |
| 134.18 | |
| Liquid |
Chemical Identifiers
| 5779-93-1 | |
| 134.178 | |
| UIFVCPMLQXKEEU-UHFFFAOYSA-N | |
| 34224 | |
| CC1=CC=CC(=C1C)C=O |
| C9H10O | |
| MFCD00798004 | |
| benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde | |
| 2,3-dimethylbenzaldehyde |
Safety and Handling
TSCA : No