missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylbenzaldehyde, 97%

$31.61 - $448.76

Chemical Identifiers

CAS 5779-93-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00798004
InChI Key UIFVCPMLQXKEEU-UHFFFAOYSA-N
Synonym benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
PubChem CID 34224
IUPAC Name 2,3-dimethylbenzaldehyde
SMILES CC1=CC=CC(=C1C)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6442506 View Documents Alfa Aesar™
H6442506
5g Each for $31.61
 
AAH6442514 View Documents Alfa Aesar™
H6442514
25g Each for $120.91
 
AAH6442522 View Documents Alfa Aesar™
H6442522
100g Each for $448.76
 
Specifications

Chemical Identifiers

5779-93-1
134.178
UIFVCPMLQXKEEU-UHFFFAOYSA-N
34224
CC1=CC=CC(=C1C)C=O
C9H10O
MFCD00798004
benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
2,3-dimethylbenzaldehyde

Specifications

1.029g/mL
86°C to 88°C (10mmHg)
5g
C9H10O
2238677
UIFVCPMLQXKEEU-UHFFFAOYSA-N
2,3-dimethylbenzaldehyde
34224
Liquid
2,3-Dimethylbenzaldehyde
1.5530
Colorless to Yellow
5779-93-1
MFCD00798004
benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
CC1=CC=CC(=C1C)C=O
134.178
134.18
97%