Please login to your online account to display your discounted pricing

2,3-Dimethylanisole, 97%, ACROS Organics™

$157.98 - $157.98

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethylanisole
CAS 2944-49-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
Formula Weight 136.19
MDL Number MFCD00008376
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC159991000

  SDS
thermo fisher scientific
159991000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC159990250

  SDS
thermo fisher scientific
159990250
Glass bottle 25g Each for $157.98
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96%
Formula Weight 136.19
Quantity 100g
Color Clear light yellow
Beilstein 06, 480
Density 0.984g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Dimethylanisole
Assay Percent Range 97%
Molecular Formula C9H12O
CAS 2944-49-2
MDL Number MFCD00008376
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5190 to 1.5220 (20°C, 589nm)
Linear Formula (CH3)2C6H3OCH3