Please login to your online account to display your discounted pricing

2,3-Dimethylanisole, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  159991000

 View more versions of this product

Catalog No. AC159991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96%
Physical Form Liquid
Formula Weight 136.19
Quantity 100g
Color Clear light yellow
Beilstein 06, 480
Density 0.984g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Dimethylanisole
Assay Percent Range 97%
Molecular Formula C9H12O
CAS 2944-49-2
MDL Number MFCD00008376
Packaging Glass bottle
Refractive Index 1.5190 to 1.5220 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H3OCH3

Specifications

Chemical Name or Material 2, 3-Dimethylanisole
CAS 2944-49-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
Formula Weight 136.19
MDL Number MFCD00008376