Please login to your online account to display your discounted pricing

2,3-Dimethylaniline, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethylaniline
Name Note 99%
CAS 87-59-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5% total base, ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C8H11N
Linear Formula (CH3)2C6H3NH2
Formula Weight 121.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115930050

  SDS
thermo fisher scientific
115930050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Assay 98.5 to 101.5% total base, ≥98.5% (GC)
Physical Form Liquid
Quantity 5mL
Color Clear yellow to red-brown
Density 0.9900g/mL
TSCA TSCA
RTECS Number ZE8750000
EINECS Number 201-755-0
Beilstein 12, 1101
Merck Index 15, 10282
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
CAS Min % 98.5
Refractive Index 1.5670 to 1.5690 (20°C, 589nm)
Infrared Spectrum Authentic
Synonym 2, 3-Xylidine
Packaging Glass bottle
Storage Information product darkens if exposed to light and air
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
MDL Number MFCD00007732
Linear Formula (CH3)2C6H3NH2
CAS 87-59-2
Name Note 99%
Assay Percent Range 99%
Chemical Name or Material 2, 3-Dimethylaniline
Molecular Formula C8H11N
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.