missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, ≥97%, Maybridge

Chemical Identifiers

CAS 910037-06-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C13H22N2O
Formula Weight 222.33
MDL Number MFCD09064998
Chemical Name or Material 2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59646CB SDS Fisher Scientific Company
CC59646CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59646DA SDS Fisher Scientific Company
CC59646DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

910037-06-8
100.0
C13H22N2O
MFCD09064998
97%
96.5
97+%
222.33
2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine

Specifications

222.33
100.0
910037-06-8
C13H22N2O
97+%
97%
250mg
Amber Glass Bottle
96.5
MFCD09064998
2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger