missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3-(Dimethylamino)propoxy]benzaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 10429-30-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD00128460
Chemical Name or Material 2-[3-(Dimethylamino)propoxy]benzaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59604DA SDS Fisher Scientific Company
CC59604DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59604DE SDS Fisher Scientific Company
CC59604DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10429-30-8
100.0
C12H17NO2
MFCD00128460
97%
96.5
97%
207.27
2-[3-(Dimethylamino)propoxy]benzaldehyde

Specifications

207.27
100.0
C12H17NO2
MFCD00128460
96.5
97%
1g
Amber Glass Bottle
97%
10429-30-8
2-[3-(Dimethylamino)propoxy]benzaldehyde

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger