Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethyl-6-nitroaniline, 98%

Chemical Identifiers

Chemical Name or Material 2,3-Dimethyl-6-nitroaniline
CAS 59146-96-2
Molecular Formula C8H10N2O2
Formula Weight 166.18g/mol
MDL Number MFCD00010148
UN Number UN2811
Synonym 6-Nitro-2,3-xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1041218

 
alfa aesar™
A10412-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Melting Point 115° to 118°C
Quantity 50g
Chemical Name or Material 2,3-Dimethyl-6-nitroaniline
CAS 59146-96-2
Assay 98%
Molecular Formula C8H10N2O2
Formula Weight 166.18g/mol
MDL Number MFCD00010148
UN Number UN2811
Synonym 6-Nitro-2,3-xylidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.