Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethyl-5-nitroindole, 97%

$328.47 - $328.47

Chemical Identifiers

Chemical Name or Material 2,3-Dimethyl-5-nitroindole
CAS 21296-94-6
Molecular Formula C10H10N2O2
Formula Weight 190.2
MDL Number MFCD00022708
Beilstein 162798
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0764406

  SDS
alfa aesar™
L0764406
5g Each for $328.47
Description & Specifications

Specifications

Formula Weight 190.2
CAS 21296-94-6
MDL Number MFCD00022708
Chemical Name or Material 2,3-Dimethyl-5-nitroindole
Molecular Formula C10H10N2O2
Assay 97%
Beilstein 162798
Quantity 5g
Melting Point 186° to 192°C