Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethyl-4-nitropyridine N-oxide, 97%

$36.64 - $120.53

Chemical Identifiers

Chemical Name or Material 2,3-Dimethyl-4-nitropyridine N-oxide
CAS 37699-43-7
Molecular Formula C7H8N2O3
Formula Weight 168.15
MDL Number MFCD00065172
Beilstein 1569438
Synonym 4-Nitro-2,3-lutidine N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945706

 
alfa aesar™
L1945706
5g Each for $120.53

AAL1945703

 
alfa aesar™
L1945703
1g Each for $36.64
Description & Specifications

Specifications

Formula Weight 168.15
Molecular Formula C7H8N2O3
MDL Number MFCD00065172
CAS 37699-43-7
Beilstein 1569438
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H302-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number UT2807000
Assay 97%
Synonym 4-Nitro-2,3-lutidine N-oxide
Chemical Name or Material 2,3-Dimethyl-4-nitropyridine N-oxide
Melting Point 92° to 94°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H341-H302-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.