Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethyl-3-hexanol, 95%

Chemical Identifiers

Chemical Name or Material 2,3-Dimethyl-3-hexanol
CAS 4166-46-5
Molecular Formula C8H18O
Formula Weight 130.23
MDL Number MFCD00046594
UN Number UN1993
Beilstein 1732656
Synonym Isopropyl methyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2452506

  SDS
alfa aesar™
B2452506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2452514

  SDS
alfa aesar™
B2452514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.23
Chemical Name or Material 2,3-Dimethyl-3-hexanol
Assay 95%
Molecular Formula C8H18O
MDL Number MFCD00046594
CAS 4166-46-5
UN Number UN1993
Beilstein 1732656
Synonym Isopropyl methyl n-propyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 49°C (120°F)
Boiling Point 157° to 159°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.