Learn More
2,3-Dimethyl-2-butanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156580050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 3-Dimethyl-2-butanol | |
97.5 | |
-14.0°C | |
0.8200g/mL | |
29°C | |
98% | |
C6H14O | |
(CH3)2CHC(CH3)2OH | |
5 mL | |
0.82 | |
IKECULIHBUCAKR-UHFFFAOYSA-N | |
2,3-dimethylbutan-2-ol | |
11670 | |
98% |
594-60-5 | |
100.0 | |
Colorless | |
120.0°C to 121.0°C | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00004463 | |
01, 413 | |
2,3-dimethyl-2-butanol, thexyl alcohol, isopropyldimethylcarbinol, 2-butanol, 2,3-dimethyl, acmc-1asco, 2,3-dimethyl-butan-2-ol, 2-butanol,2,3-dimethyl, ikeculihbucakr-uhfffaoysa, ch3 2chc oh ch3 2 | |
CC(C)C(C)(C)O | |
102.18 | |
102.18 | |
Liquid |
Chemical Identifiers
594-60-5 | |
102.18 | |
IKECULIHBUCAKR-UHFFFAOYSA-N | |
11670 | |
CC(C)C(C)(C)O |
C6H14O | |
MFCD00004463 | |
2,3-dimethyl-2-butanol, thexyl alcohol, isopropyldimethylcarbinol, 2-butanol, 2,3-dimethyl, acmc-1asco, 2,3-dimethyl-butan-2-ol, 2-butanol,2,3-dimethyl, ikeculihbucakr-uhfffaoysa, ch3 2chc oh ch3 2 | |
2,3-dimethylbutan-2-ol |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-847-2
RUO – Research Use Only