Please login to your online account to display your discounted pricing

2,3-dimethyl-1h-indole-5-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H11NO2
Formula Weight 189.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07746CB

  SDS
fisher scientific company
MO07746CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.21
Quantity 250mg
Assay 97%
CAS 14844-73-6
Assay Percent Range 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Molecular Formula C11H11NO2