Please login to your online account to display your discounted pricing

2,3-dimethyl-1h-indole-5-carboxylic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO07746CB

 View more versions of this product

Catalog No. MO07746CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 189.21
Quantity 250mg
Assay 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Assay Percent Range 97%
CAS 14844-73-6
Molecular Formula C11H11NO2

Specifications

Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H11NO2
Formula Weight 189.21