Learn More
2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0420718
Description
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
2,3-Dimethyl-1,3-butadiene | |
513-81-5 | |
0.726 | |
−1°C (30°F) | |
1.438 | |
50 g | |
605285 | |
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene | |
SDJHPPZKZZWAKF-UHFFFAOYSA-N | |
2,3-dimethylbuta-1,3-diene | |
10566 | |
98% |
stabilized with 100ppm BHT | |
-76°C | |
68°C to 69°C | |
C6H10 | |
MFCD00008595 | |
UN3295 | |
14,3238 | |
Miscible with chloroform. Immiscible with water. | |
CC(=C)C(C)=C | |
82.15 | |
82.15 |
Chemical Identifiers
513-81-5 | |
82.15 | |
SDJHPPZKZZWAKF-UHFFFAOYSA-N | |
10566 | |
CC(=C)C(C)=C |
C6H10 | |
MFCD00008595 | |
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene | |
2,3-dimethylbuta-1,3-diene |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 208-172-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only