Please login to your online account to display your discounted pricing

2,3-Dimethoxyphenethylamine, 97%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC439710010

 View more versions of this product

Catalog No. AC439710010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 181.23
Quantity 1g
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H15NO2
CAS 3213-29-4
Chemical Name or Material 2, 3-Dimethoxyphenethylamine

Specifications

Chemical Name or Material 2, 3-Dimethoxyphenethylamine
Assay Percent Range 97%
Molecular Formula C10H15NO2
Formula Weight 181.23