Please login to your online account to display your discounted pricing

2,3-Dimethoxyphenethylamine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethoxyphenethylamine
Assay Percent Range 97%
Molecular Formula C10H15NO2
Formula Weight 181.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC439710050

  SDS
thermo fisher scientific
AC439710050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439710010

  SDS
thermo fisher scientific
AC439710010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2, 3-Dimethoxyphenethylamine
Quantity 5g
CAS 3213-29-4
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H15NO2
Formula Weight 181.23