missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxynaphthalene, 97%

Manufacturer:  Alfa Aesar™ H5552303

 View more versions of this product

Catalog No. AAH5552303



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Notes
Refrigerated (2-4°C). Keep away from oxidizing agents.
Specifications

Specifications

2,3-Dimethoxynaphthalene
116°C to 121°C
10103-06-7
MFCD05664308
XYRPWXOJBNGTMX-UHFFFAOYSA-N
2,3-dimethoxynaphthalene
604708
Powder
White to Yellow
1g
C12H12O2
2,3-dihydroxynaphthalene, dimethyl ether
COC1=CC2=CC=CC=C2C=C1OC
188.226
188.23
97%

Chemical Identifiers

10103-06-7
188.226
XYRPWXOJBNGTMX-UHFFFAOYSA-N
604708
COC1=CC2=CC=CC=C2C=C1OC
C12H12O2
MFCD05664308
2,3-dihydroxynaphthalene, dimethyl ether
2,3-dimethoxynaphthalene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents