Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxynaphthalene, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Dimethoxynaphthalene
CAS 10103-06-7
Formula Weight 188.23
MDL Number MFCD05664308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5552303

  SDS
alfa aesar™
H5552303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5552306

  SDS
alfa aesar™
H5552306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5552314

  SDS
alfa aesar™
H5552314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.23
Melting Point 116° to 121°C
MDL Number MFCD05664308
Physical Form Powder
CAS 10103-06-7
Quantity 1g
Assay 97%
Chemical Name or Material 2,3-Dimethoxynaphthalene
Color White to yellow

Applications
It is used as a pharmaceutical intermediate.

Notes
Refrigerated (2-4°C). Keep away from oxidizing agents.