missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dimethoxybenzylamine, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC433390250

 View more versions of this product

Catalog No. AC433390250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3-Dimethoxybenzylamine
113°C
Glass bottle
140.0°C to 142.0°C (15.0mmHg)
25g
98.5
99%
MFCD00052392
OHKBVLWPESSWKC-UHFFFAOYSA-N
4-[(4-chlorophenyl)methyl]pyridine
78124
Liquid
1.1100g/mL
Authentic
1.11
Colorless to Yellow
4409-11-4
100.0
C12H10ClN
4-4-chlorobenzyl pyridine, 4-4-chlorophenyl methyl pyridine, 4-4'-chlorobenzyl pyridine, 4-4-chloro-benzyl-pyridine, 4-para-chlorobenzyl-pyridine, 4-4-chlorophenyl methyl pyridine, pyridine, 4-4-chlorophenyl methyl, pyridine, 4-4-chlorophenyl methyl, acmc-2097fd, 4-p-chlorobenzyl pyridine
C1=CC(=CC=C1CC2=CC=NC=C2)Cl
203.67
167.21
99%

Chemical Identifiers

4409-11-4
203.67
OHKBVLWPESSWKC-UHFFFAOYSA-N
78124
C1=CC(=CC=C1CC2=CC=NC=C2)Cl
C12H10ClN
MFCD00052392
4-4-chlorobenzyl pyridine, 4-4-chlorophenyl methyl pyridine, 4-4'-chlorobenzyl pyridine, 4-4-chloro-benzyl-pyridine, 4-para-chlorobenzyl-pyridine, 4-4-chlorophenyl methyl pyridine, pyridine, 4-4-chlorophenyl methyl, pyridine, 4-4-chlorophenyl methyl, acmc-2097fd, 4-p-chlorobenzyl pyridine
4-[(4-chlorophenyl)methyl]pyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents