Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxybenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2,3-Dimethoxybenzonitrile
CAS 5653-62-3
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00001784
UN Number UN3439
Beilstein 2719631
Synonym 3-Cyanoveratrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2094209

  SDS
alfa aesar™
B2094209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2094218

  SDS
alfa aesar™
B2094218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.18
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H9NO2
MDL Number MFCD00001784
CAS 5653-62-3
UN Number UN3439
Quantity 10g
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 44° to 47°C
Beilstein 2719631
Synonym 3-Cyanoveratrole
RTECS Number DI4355000
Chemical Name or Material 2,3-Dimethoxybenzonitrile
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%

Applications
It is employed in the replacement of nuclear alkoxyl groups to yield dimethoxy ketones by the action of methyl and phenyl Grignard reagents on 2,3-dimethoxybenzonitrile.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.