missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dimethoxybenzoic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC115421000
Specifications
2, 3-Dimethoxybenzoic acid | |
Glass Bottle | |
121°C to 124°C | |
1521-38-6 | |
100.0 | |
C9H10O4 | |
MFCD00002432 | |
2,3-dimethoxy benzoic acid | |
FODBVCSYJKNBLO-UHFFFAOYSA-N | |
2,3-dimethoxybenzoic acid | |
15204 | |
Fine Crystalline Powder |
Authentic | |
Beige | |
100g | |
98.5 | |
98.5% min. (HPLC) | |
(CH3O)2C6H3CO2H | |
10, I, 174 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol | |
COC1=CC=CC(=C1OC)C(=O)O | |
182.18 | |
182.18 | |
99% |
Chemical Identifiers
1521-38-6 | |
182.18 | |
FODBVCSYJKNBLO-UHFFFAOYSA-N | |
15204 | |
COC1=CC=CC(=C1OC)C(=O)O |
C9H10O4 | |
MFCD00002432 | |
2,3-dimethoxy benzoic acid | |
2,3-dimethoxybenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 216-188-4