missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxybenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1459230

 View more versions of this product

Catalog No. AAA1459230



Specifications

Specifications

2,3-Dimethoxybenzoic acid
121°C to 124°C
1521-38-6
MFCD00002432
o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
COC1=CC=CC(=C1OC)C(=O)O
182.175
182.18
Odorless
250g
C9H10O4
2210858
FODBVCSYJKNBLO-UHFFFAOYSA-N
2,3-dimethoxybenzoic acid
15204
99%

Chemical Identifiers

1521-38-6
182.175
FODBVCSYJKNBLO-UHFFFAOYSA-N
15204
COC1=CC=CC(=C1OC)C(=O)O
C9H10O4
MFCD00002432
o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
2,3-dimethoxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DG8598700

Documents