Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxybenzaldoxime, 97%

$33.86 - $103.14

Chemical Identifiers

Chemical Name or Material 2,3-Dimethoxybenzaldoxime
CAS 5470-95-1
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00033234
Beilstein 1953084
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1377603

 
alfa aesar™
L1377603
1g Each for $33.86

AAL1377606

 
alfa aesar™
L1377606
5g Each for $103.14
Description & Specifications

Specifications

Formula Weight 181.19
Chemical Name or Material 2,3-Dimethoxybenzaldoxime
MDL Number MFCD00033234
CAS 5470-95-1
Assay 97%
Molecular Formula C9H11NO3
Beilstein 1953084
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number YX5089710
Melting Point 98° to 100°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.