missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxybenzaldoxime, 97%

Manufacturer:  Alfa Aesar™ L1377603

 View more versions of this product

Catalog No. AAL1377603



Specifications

Specifications

2,3-Dimethoxybenzaldoxime
1g
C9H11NO3
1953084
GKAIQNACVNFHCU-UXBLZVDNSA-N
(NE)-N-[(2,3-dimethoxyphenyl)methylidene]hydroxylamine
6871292
97%
98°C to 100°C
5470-95-1
MFCD00033234
2,3-dimethoxybenzaldoxime, o-veratraldehyde, oxime, benzaldehyde, 2,3-dimethoxy-, oxime, e-n-2,3-dimethoxyphenyl methylidene hydroxylamine, ne-n-2,3-dimethoxyphenyl methylidene hydroxylamine, z-2,3-dimethoxybenzaldehyde oxime, benzaldehyde, 2,3-dimethoxy-,oxime, 2-08-00-00269 beilstein handbook reference, 1e-2,3-dimethoxybenzaldehyde oxime, 2,3-dimethoxyphenyl hydroxyimino methane
COC1=CC=CC(=C1OC)C=NO
181.191
181.19

Chemical Identifiers

5470-95-1
181.191
GKAIQNACVNFHCU-UXBLZVDNSA-N
6871292
COC1=CC=CC(=C1OC)C=NO
C9H11NO3
MFCD00033234
2,3-dimethoxybenzaldoxime, o-veratraldehyde, oxime, benzaldehyde, 2,3-dimethoxy-, oxime, e-n-2,3-dimethoxyphenyl methylidene hydroxylamine, ne-n-2,3-dimethoxyphenyl methylidene hydroxylamine, z-2,3-dimethoxybenzaldehyde oxime, benzaldehyde, 2,3-dimethoxy-,oxime, 2-08-00-00269 beilstein handbook reference, 1e-2,3-dimethoxybenzaldehyde oxime, 2,3-dimethoxyphenyl hydroxyimino methane
(NE)-N-[(2,3-dimethoxyphenyl)methylidene]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YX5089710

Documents