missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442200050

 View more versions of this product

Catalog No. AC442200050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 3-Dimethoxy-5-methyl-1, 4-benzoquinone
Red to Orange
5g
97.5
97.5% min. (HPLC)
2,3-dimethoxy-5-methyl-1,4-benzoquinone, 2,3-dimethoxy-5-methyl-p-benzoquinone, 2,3-dimethoxy-5-methylbenzoquinone, 2-methyl-4,5-dimethoxy-p-quinone, coenzyme q0, coq0, ubiquinone 0, ubiquinone q0, ubiquinone-0, ubiquinone-o
CC1=CC(=O)C(=C(C1=O)OC)OC
182.18
CHEBI:27906
Crystalline Powder or Needles
Authentic
59°C to 61°C
605-94-7
100.0
C9H10O4
UIXPTCZPFCVOQF-UHFFFAOYSA-N
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
69068
182.18
98%

Chemical Identifiers

605-94-7
182.18
2,3-dimethoxy-5-methyl-1,4-benzoquinone, 2,3-dimethoxy-5-methyl-p-benzoquinone, 2,3-dimethoxy-5-methylbenzoquinone, 2-methyl-4,5-dimethoxy-p-quinone, coenzyme q0, coq0, ubiquinone 0, ubiquinone q0, ubiquinone-0, ubiquinone-o
CHEBI:27906
CC1=CC(=O)C(=C(C1=O)OC)OC
C9H10O4
UIXPTCZPFCVOQF-UHFFFAOYSA-N
69068
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-100-8

SDS Documents