Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%

$189.85 - $763.93

Chemical Identifiers

Chemical Name or Material 2,3-Dimethoxy-5-methyl-1,4-benzoquinone
CAS 605-94-7
Molecular Formula C9H10O4
Formula Weight 182.18
MDL Number MFCD00001595
Beilstein 1640422
Synonym Coenzyme Q0
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2477706

  SDS
alfa aesar™
B2477706
5g Each for $189.85

AAB2477714

  SDS
alfa aesar™
B2477714
25g Each for $763.93
Description & Specifications

Specifications

Formula Weight 182.18
Molecular Formula C9H10O4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Beilstein 1640422
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,3-Dimethoxy-5-methyl-1,4-benzoquinone
Quantity 5g
Synonym Coenzyme Q0
MDL Number MFCD00001595
Health Hazard 1 Warning
Melting Point 57° to 60°C
CAS 605-94-7
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.