Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%

$70.35 - $193.95

Chemical Identifiers

Chemical Name or Material 2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate
CAS 207233-91-8
Molecular Formula C3H7NaO3S3·H2O
Formula Weight 228.30 (210.28 Anhydrous)
MDL Number MFCD00149543
Beilstein 3734863
Merck Index 14,3210
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1951903

 
alfa aesar™
L1951903
1g Each for $193.95

AAL19519MD

 
alfa aesar™
L19519MD
250mg Each for $70.35
Description & Specifications

Specifications

Formula Weight 228.30 (210.28 Anhydrous)
Chemical Name or Material 2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate
Molecular Formula C3H7NaO3S3·H2O
CAS 207233-91-8
Quantity 1g
MDL Number MFCD00149543
Merck Index 14,3210
Assay 95%
Melting Point ≈229°C
Beilstein 3734863
RTECS Number TZ6420000

Applications
Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

Solubility
Soluble in Water: 100 mg/ml

Notes
Store at 2-8°C. Incompatible with oxidizing agents.