missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydroxynaphthalene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115022500

 View more versions of this product

Catalog No. AC115022500



Description

Description

Specifications

Specifications

2, 3-Dihydroxynaphthalene
Authentic
Beige-Gray to White
250g
92-44-4
100.0
C10H8O2
MFCD00004073
2,3-dihydroxy naphthalene, 2,3-dihydroxylnaphthalene, 2,3-dihydroxynaphthalene, 2,3-dihydroxynaphthlene, 2,3-dihydroxynapthalene, 2,3-naphthalenediol, 2,3dihydroxynaphthalene, 3-hydroxy-2-naphthol, acmc-209rfr, naphthalenediol-2,3
JRNGUTKWMSBIBF-UHFFFAOYSA-N
naphthalene-2,3-diol
7091
160.17
97%
175°C
Plastic bottle
162.0°C to 166.0°C
98%
97.5
96% min. (HPLC)
C10H6(OH)2
06, 982
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
C1=CC=C2C=C(C(=CC2=C1)O)O
160.17
CHEBI:38135
Powder

Chemical Identifiers

92-44-4
160.17
JRNGUTKWMSBIBF-UHFFFAOYSA-N
7091
naphthalene-2,3-diol
C10H8O2
MFCD00004073
2,3-dihydroxy naphthalene, 2,3-dihydroxylnaphthalene, 2,3-dihydroxynaphthalene, 2,3-dihydroxynaphthlene, 2,3-dihydroxynapthalene, 2,3-naphthalenediol, 2,3dihydroxynaphthalene, 3-hydroxy-2-naphthol, acmc-209rfr, naphthalenediol-2,3
CHEBI:38135
C1=CC=C2C=C(C(=CC2=C1)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-156-7

RTECSNumber : QJ4750000

TSCA : TSCA

SDS Documents