missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',3-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2784103

 View more versions of this product

Catalog No. AAH2784103



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2',3-Dihydroxyflavone
201°C to 204°C
MFCD00017674
3,2'-dihydroxyflavone, 2',3-dihydroxyflavone, 3-hydroxy-2-2-hydroxyphenyl-4h-chromen-4-one, 2'-hydroxyflavonol, 3-hydroxy-2-2-hydroxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-2-hydroxyphenyl, 2'3-dioh-flavone, 3,2'-dihydroxy flavone, acmc-1b19t, 7,8-dihydroxyflavone-derivative-1
OC1=CC=CC=C1C1=C(O)C(=O)C2=CC=CC=C2O1
254.24
254.24
6068-76-4
C15H10O4
1 g
VECGDSZOFMYGAF-UHFFFAOYSA-N
3-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
455313
97%

Chemical Identifiers

6068-76-4
254.24
VECGDSZOFMYGAF-UHFFFAOYSA-N
455313
OC1=CC=CC=C1C1=C(O)C(=O)C2=CC=CC=C2O1
C15H10O4
MFCD00017674
3,2'-dihydroxyflavone, 2',3-dihydroxyflavone, 3-hydroxy-2-2-hydroxyphenyl-4h-chromen-4-one, 2'-hydroxyflavonol, 3-hydroxy-2-2-hydroxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-2-hydroxyphenyl, 2'3-dioh-flavone, 3,2'-dihydroxy flavone, acmc-1b19t, 7,8-dihydroxyflavone-derivative-1
3-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only