missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydroxybenzoic acid, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 155170250

 View more versions of this product

Catalog No. AC155170250



Specifications

Specifications

2, 3-Dihydroxybenzoic acid
Glass bottle
204°C to 208°C
99%
98.5
99%
(HO)2C6H3CO2H
10, 375
GLDQAMYCGOIJDV-UHFFFAOYSA-N
2,3-dihydroxybenzoic acid
19
154.12
99%
Authentic
Beige to Brown or Gray-Pink
25g
303-38-8
100.0
C7H6O4
MFCD00002446
2,3-dihydroxy-benzoic acid
C1=CC(=C(C(=C1)O)O)C(=O)O
154.12
CHEBI:18026
Crystalline Powder

Chemical Identifiers

303-38-8
154.12
GLDQAMYCGOIJDV-UHFFFAOYSA-N
19
2,3-dihydroxybenzoic acid
C7H6O4
MFCD00002446
2,3-dihydroxy-benzoic acid
CHEBI:18026
C1=CC(=C(C(=C1)O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 206-139-5

RTECSNumber : DG8576490

TSCA : TSCA

SDS Documents