missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydropyrido[2,3-d]oxazol-2-one, 97%, Alfa Aesar™

$131.16 - $577.62

Chemical Identifiers

CAS 60832-72-6
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD09065038
InChI Key OVLXOTUWFLHWQT-UHFFFAOYSA-N
Synonym 1,3 oxazolo 4,5-b pyridin-2 3h-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 2-oxooxazolo 4,5-b pyridine, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, pubchem11061
PubChem CID 2799900
ChEBI CHEBI:38580
IUPAC Name 3H-[1,3]oxazolo[4,5-b]pyridin-2-one
SMILES C1=CC2=C(NC(=O)O2)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6623414 View Documents Thermo Fisher Scientific
H6623414
25g Each for $131.16
 
AAH6623422 View Documents Thermo Fisher Scientific
H6623422
100g Each for $577.62
 
Specifications

Chemical Identifiers

60832-72-6
136.11
OVLXOTUWFLHWQT-UHFFFAOYSA-N
2799900
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
C6H4N2O2
MFCD09065038
1,3 oxazolo 4,5-b pyridin-2 3h-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 2-oxooxazolo 4,5-b pyridine, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, pubchem11061
CHEBI:38580
C1=CC2=C(NC(=O)O2)N=C1

Specifications

25g
C6H4N2O2
125648
OVLXOTUWFLHWQT-UHFFFAOYSA-N
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
2799900
136.11
97%
60832-72-6
MFCD09065038
1,3 oxazolo 4,5-b pyridin-2 3h-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 2-oxooxazolo 4,5-b pyridine, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, pubchem11061
C1=CC2=C(NC(=O)O2)N=C1
136.11
CHEBI:38580
Solid
2,3-Dihydropyrido[2,3-d]oxazol-2-one
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures