missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydropyrido[2,3-d]oxazol-2-one, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H6623422

 View more versions of this product

Catalog No. AAH6623422



Specifications

Specifications

2,3-Dihydropyrido[2,3-d]oxazol-2-one
60832-72-6
MFCD09065038
1,3 oxazolo 4,5-b pyridin-2 3h-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 2-oxooxazolo 4,5-b pyridine, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, pubchem11061
C1=CC2=C(NC(=O)O2)N=C1
136.11
CHEBI:38580
Solid
100g
C6H4N2O2
125648
OVLXOTUWFLHWQT-UHFFFAOYSA-N
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
2799900
136.11
97%

Chemical Identifiers

60832-72-6
136.11
OVLXOTUWFLHWQT-UHFFFAOYSA-N
2799900
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
C6H4N2O2
MFCD09065038
1,3 oxazolo 4,5-b pyridin-2 3h-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 2-oxooxazolo 4,5-b pyridine, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 3h-oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, pubchem11061
CHEBI:38580
C1=CC2=C(NC(=O)O2)N=C1
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents