Learn More
2,3-Dihydrofuran, 98+%, Thermo Scientific Chemicals
$61.27 - $475.63
Chemical Identifiers
CAS | 1191-99-7 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00003205 |
InChI Key | JKTCBAGSMQIFNL-UHFFFAOYSA-N |
PubChem CID | 70934 |
ChEBI | CHEBI:51662 |
IUPAC Name | 2,3-dihydrofuran |
SMILES | C1CC=CO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2057514
|
Thermo Scientific Chemicals
B2057514 |
25 g |
Each for $61.27
|
|
|||||
AAB2057522
|
Thermo Scientific Chemicals
B2057522 |
100 g |
Each for $116.08
|
|
|||||
AAB2057536
|
Thermo Scientific Chemicals
B2057536 |
500 g |
Each for $475.63
|
|
|||||
Description
2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids. It is applied for alkylation by active methylene compounds, catalyzed by trans-Dichlorobis(triphenylÂphosphine)Âpalladium(II). It is also used in the preparation of biologically active compounds such as antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids. It is applied for alkylation by active methylene compounds, catalyzed by trans-Dichlorobis(triphenylÂphosphine)Âpalladium(II). It is also used in the preparation of biologically active compounds such as antitumor agents.
Solubility
Soluble in water at 20°C, 20g/L and chloroform.
Notes
Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
Chemical Identifiers
1191-99-7 | |
70.09 | |
JKTCBAGSMQIFNL-UHFFFAOYSA-N | |
CHEBI:51662 | |
C1CC=CO1 |
C4H6O | |
MFCD00003205 | |
70934 | |
2,3-dihydrofuran |
Specifications
1191-99-7 | |
54°C to 55°C | |
Pungent | |
1.4245 | |
25 g | |
103168 | |
Soluble in water at 20°C,20g/L and chloroform. | |
C1CC=CO1 | |
70.09 | |
CHEBI:51662 | |
≥98% |
0.927 | |
−24°C (−11°F) | |
C4H6O | |
MFCD00003205 | |
UN3271 | |
4,5-dihydrofuran, furan, 2,3-dihydro, furan, dihydro, dihydrofuran, 2,3 dihydrofuran, dihydrofurane, 2,3dihydrofuran, furan,3-dihydro, 2,3-dihydrofurane, 2,3-dihydro-furan | |
JKTCBAGSMQIFNL-UHFFFAOYSA-N | |
2,3-dihydrofuran | |
70934 | |
70.09 | |
2,3-Dihydrofuran |
Safety and Handling
GHS H Statement
H225-H301-H319
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P372-P374-P380-P501c
H225-H302-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 214-747-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only