Please login to your online account to display your discounted pricing

2,3-Dihydrobenzofuran, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dihydrobenzofuran
Name Note 98%
CAS 496-16-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C8H8O
Formula Weight 120.15g/mol
MDL Number MFCD00005855
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179940050

  SDS
thermo fisher scientific
179940050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
Water 0.1% max.
Density 1.06g/mL
Color Clear colorless to light yellow
Quantity 5g
Chemical Name or Material 2, 3-Dihydrobenzofuran
Name Note 98%
CAS 496-16-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear colorless to light yellow liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C8H8O
Formula Weight 120.15g/mol
MDL Number MFCD00005855
Beilstein 17, 50
Merck Index 15, 2547
Synonym Coumaran
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 207-817-3
TSCA TSCA
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.