Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dihydrobenzo[b]furan, 98%

$73.57 - $238.59

Chemical Identifiers

Chemical Name or Material 2,3-Dihydrobenzo[b]furan
CAS 496-16-2
Molecular Formula C8H8O
Formula Weight 120.15
MDL Number MFCD00005855
Beilstein 111928
Merck Index 14,2559
Synonym Coumaran; 2,3-Dihydro-2,3-benzofuran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1136806

 
alfa aesar™
A1136806
5g Each for $73.57

AAA1136814

 
alfa aesar™
A1136814
25g Each for $238.59
Description & Specifications

Specifications

Formula Weight 120.15
Chemical Name or Material 2,3-Dihydrobenzo[b]furan
TSCA TSCA
Assay 98%
Molecular Formula C8H8O
MDL Number MFCD00005855
Melting Point −21°C
CAS 496-16-2
Beilstein 111928
Merck Index 14,2559
Synonym Coumaran; 2,3-Dihydro-2,3-benzofuran
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 66°C (150°F)
Boiling Point 188° to 189°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.