missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dihydrobenzo[b]furan, 98%

Manufacturer:  Alfa Aesar™ A1136822

 View more versions of this product

Catalog No. AAA1136822


 

Specifications

Specifications

2,3-Dihydrobenzo[b]furan
188° to 189°C
496-16-2
C8H8O
−21°C
14,2559
HBEDSQVIWPRPAY-UHFFFAOYSA-N
2,3-dihydro-1-benzofuran
10329
120.15
66°C (150°F)
100g
98%
MFCD00005855
111928
Coumaran; 2,3-Dihydro-2,3-benzofuran
C1COC2=CC=CC=C21
120.151
CHEBI:87607

Chemical Identifiers

496-16-2
120.151
HBEDSQVIWPRPAY-UHFFFAOYSA-N
10329
C8H8O
MFCD00005855
Coumaran; 2,3-Dihydro-2,3-benzofuran
CHEBI:87607
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents