missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-7-carboxylic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC00901DA

 View more versions of this product

Catalog No. CC00901DA



Specifications

Specifications

2, 3-Dihydrobenzo[b]furan-7-carboxylic acid
5g
96.5
97%
MFCD00671526
UHXBMSNEECJPSX-UHFFFAOYSA-N
2,3-dihydro-1-benzofuran-7-carboxylic acid
2795014
97%
Amber Glass Bottle
35700-40-4
100.0
C9H8O3
2,3-dihydrobenzofuran-7-carboxylic acid, 2,3-dihydrobenzo b furan-7-carboxylic acid, 7-coumarancarboxylic acid, 2,3-dihydro-7-benzofurancarboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro, 7-benzofurancarboxylic acid, 2,3-dihydro, pubchem16734, acmc-209iiq, dihydrobenzofurancarboxylicacid, 7-dihydrobenzofuran carboxylic acid
C1COC2=C(C=CC=C21)C(=O)O
164.16
164.16

Chemical Identifiers

35700-40-4
164.16
UHXBMSNEECJPSX-UHFFFAOYSA-N
2795014
C1COC2=C(C=CC=C21)C(=O)O
C9H8O3
MFCD00671526
2,3-dihydrobenzofuran-7-carboxylic acid, 2,3-dihydrobenzo b furan-7-carboxylic acid, 7-coumarancarboxylic acid, 2,3-dihydro-7-benzofurancarboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro, 7-benzofurancarboxylic acid, 2,3-dihydro, pubchem16734, acmc-209iiq, dihydrobenzofurancarboxylicacid, 7-dihydrobenzofuran carboxylic acid
2,3-dihydro-1-benzofuran-7-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents