missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00801EA
Specifications
2, 3-Dihydrobenzo[b]furan-5-carboxylic acid | |
97% | |
C9H8O3 | |
2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan | |
C1COC2=C1C=C(C=C2)C(=O)O | |
164.16 | |
164.16 |
76429-73-7 | |
Amber Glass Bottle | |
10g | |
YXYOLVAXVPOIMA-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-5-carboxylic acid | |
676583 | |
97% |
Chemical Identifiers
76429-73-7 | |
164.16 | |
2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan | |
2,3-dihydro-1-benzofuran-5-carboxylic acid |
C9H8O3 | |
YXYOLVAXVPOIMA-UHFFFAOYSA-N | |
676583 | |
C1COC2=C1C=C(C=C2)C(=O)O |