Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%


Quantity 25g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC00801EB

 View more versions of this product

Catalog No. CC00801EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 164.16
Quantity 25g
Assay 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8O3
CAS 76429-73-7

Specifications

Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8O3
Formula Weight 164.16