Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-5-carbothioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbothioamide
CAS 306936-08-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NOS
Formula Weight 179.24
MDL Number MFCD00728868
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00108CB

 
fisher scientific company
SP00108CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.24
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbothioamide
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H9NOS
MDL Number MFCD00728868
CAS 306936-08-3