Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-5-carbothioamide, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SP00108CB

 View more versions of this product

Catalog No. SP00108CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 179.24
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbothioamide
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H9NOS
MDL Number MFCD00728868
CAS 306936-08-3

Specifications

Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbothioamide
CAS 306936-08-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NOS
Formula Weight 179.24
MDL Number MFCD00728868