missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydrobenzo[b]furan-5-carbaldehyde, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00804DA
Specifications
2, 3-Dihydrobenzo[b]furan-5-carbaldehyde | |
94.5 | |
95% | |
C9H8O2 | |
1g | |
WEBVDBDZSOJGPB-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-5-carbaldehyde | |
735901 | |
95% |
55745-70-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD00068058 | |
2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde | |
O=CC1=CC=C2OCCC2=C1 | |
148.16 | |
148.16 |