Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-5-carbaldehyde, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC00804CB

 View more versions of this product

Catalog No. CC00804CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 148.16
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbaldehyde
Packaging Amber Glass Bottle
Molecular Formula C9H8O2
MDL Number MFCD00068058
CAS Min % 94.5
Assay Percent Range 95%
CAS 55745-70-5

Specifications

Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbaldehyde
CAS 55745-70-5
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H8O2
Formula Weight 148.16
MDL Number MFCD00068058